Thermo Scientific Chemicals

4'-Iodoacetophenone, 98%, Thermo Scientific Chemicals

Catalog number: 278890250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

4'-Iodoacetophenone, 98%, Thermo Scientific Chemicals

Catalog number: 278890250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 278890250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
13329-40-3
IUPAC Name
1-(4-iodophenyl)ethan-1-one
Molecular Formula
C8H7IO
InChI Key
JZJWCDQGIPQBAO-UHFFFAOYSA-N
SMILES
CC(=O)C1=CC=C(I)C=C1
Appearance (Color)
Yellow to brown
Appearance (Form)
Crystalline powder
GC
>=97.5 %
Melting point
82°C to 89°C
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text