2-chlorobenzyl bromide, 97%, Thermo Scientific Chemicals
2-chlorobenzyl bromide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-chlorobenzyl bromide, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 278930050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS611-17-6
IUPAC Name1-(bromomethyl)-2-chlorobenzene
Molecular FormulaC7H6BrCl
InChI KeyPURSZYWBIQIANP-UHFFFAOYSA-N
SMILESClC1=CC=CC=C1CBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.5910 to 1.5930 (20°C, 589 nm)
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only