Thermo Scientific Chemicals

2-chlorobenzyl bromide, 97%, Thermo Scientific Chemicals

Catalog number: 278930250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

2-chlorobenzyl bromide, 97%, Thermo Scientific Chemicals

Catalog number: 278930250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 278930250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
611-17-6
IUPAC Name
1-(bromomethyl)-2-chlorobenzene
Molecular Formula
C7H6BrCl
InChI Key
PURSZYWBIQIANP-UHFFFAOYSA-N
SMILES
ClC1=CC=CC=C1CBr
Appearance (Color)
Clear yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Refractive index
1.5910 to 1.5930 (20°C, 589 nm)
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text