Thermo Scientific Chemicals

Silver sulfate, pure, 10 g/l solution in sulfuric acid, Thermo Scientific Chemicals

Catalog number: 279480010
1 L, Glass bottle, Each
Thermo Scientific Chemicals

Silver sulfate, pure, 10 g/l solution in sulfuric acid, Thermo Scientific Chemicals

Catalog number: 279480010
1 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 279480010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10294-26-5, 7664-93-9
Appearance (Color)
Clear colorless
Solution contains
9.5 to 10.5 g/l Ag2SO4 in
Solution contains
H2SO4 95-97%
Appearance (Form)
Solution

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text