Thermo Scientific Chemicals

Zeolite, type: ZSM-5, Thermo Scientific Chemicals

Catalog number: 279571000
100 g, Each
Thermo Scientific Chemicals

Zeolite, type: ZSM-5, Thermo Scientific Chemicals

Catalog number: 279571000
100 g, Each
Quantity
Catalog number: 279571000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
308081-08-5
IUPAC Name
5-(2-ethyl-2,3-dihydro-1H-inden-2-yl)-1H-imidazole
Molecular Formula
C14H16N2
InChI Key
HSWPZIDYAHLZDD-UHFFFAOYSA-N
SMILES
CCC1(CC2=CC=CC=C2C1)C1=CN=CN1
Appearance (Color)
White to beige to light brown
Composition
400 to 570 Molar ratio SiO2/Al2O3
Loss on ignition
=<10 % (1000°C)
Appearance (Form)
Powder
Sodium (Na)
=<700 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text