Thermo Scientific Chemicals

2-Aminobenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: 279660010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Aminobenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: 279660010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 279660010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
529-23-7
IUPAC Name
2-aminobenzaldehyde
Molecular Formula
C7H7NO
InChI Key
FXWFZIRWWNPPOV-UHFFFAOYSA-N
SMILES
NC1=CC=CC=C1C=O
Appearance (Color)
Yellow
Melting point
34°C to 40°C
Appearance (Form)
Powder or crystals or flakes
Infrared spectrum
Conforms
HPLC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text