Thermo Scientific Chemicals

Biphenylene, 99+%, Thermo Scientific Chemicals

Catalog number: 279742500
250 mg, Glass bottle, Each
Thermo Scientific Chemicals

Biphenylene, 99+%, Thermo Scientific Chemicals

Catalog number: 279742500
250 mg, Glass bottle, Each
Quantity
Packaging
Catalog number: 279742500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
259-79-0
IUPAC Name
biphenylene
Molecular Formula
C12H8
InChI Key
IFVTZJHWGZSXFD-UHFFFAOYSA-N
SMILES
C1=CC=C2C(=C1)C1=CC=CC=C21
Appearance (Form)
Needles
Melting point
109°C to 112°C
Appearance (Color)
Yellow
Infrared spectrum
Conforms
GC
>=99.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text