Methyl cinnamate, 98%, Thermo Scientific Chemicals
Methyl cinnamate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl cinnamate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
Packaging:
Glass bottle
Catalog number 279765000
Price (USD)/ Each
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS103-26-4
IUPAC Namemethyl (2E)-3-phenylprop-2-enoate
Molecular FormulaC10H10O2
InChI KeyCCRCUPLGCSFEDV-BQYQJAHWSA-N
SMILESCOC(=O)\C=C\C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Fused crystalline mass
Melting point33.5°C to 36°C
Infrared spectrumConforms
GC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only