2-Chloro-2-ethoxyacetic acid ethyl ester, 90%, remainder mainly ethyl diethoxyacetat, Thermo Scientific Chemicals
2-Chloro-2-ethoxyacetic acid ethyl ester, 90%, remainder mainly ethyl diethoxyacetat, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Chloro-2-ethoxyacetic acid ethyl ester, 90%, remainder mainly ethyl diethoxyacetat, Thermo Scientific Chemicals

Have Questions?
Quantity:
50 g
Catalog number 279870500
Price (USD)/ Each
-
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS34006-60-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to orange-brown
Appearance (Form)Liquid
GC>=88 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only