2-Methyl-5-tert-butylthiophenol, 93%, remainder isomer, Thermo Scientific Chemicals
2-Methyl-5-tert-butylthiophenol, 93%, remainder isomer, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Methyl-5-tert-butylthiophenol, 93%, remainder isomer, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
50 g
Packaging:
Glass bottle
Catalog number 279960500
Price (USD)/ Each
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7340-90-1
IUPAC Name(5-tert-butyl-2-methylphenyl)sulfanide
Molecular FormulaC11H15S
InChI KeyZSBNXOIAJFVXMP-UHFFFAOYSA-M
SMILESCC1=CC=C(C=C1[S-])C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=91 %
Infrared spectrumConforms
Refractive index1.5460 to 1.5480 (20°C, 589 nm)
Appearance (Form)Liquid
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only