Thermo Scientific™

Pierce™ Biotin Quantitation Kit

Catalog number: 28005
Have Questions?
Thermo Scientific™

Pierce™ Biotin Quantitation Kit

Catalog number: 28005
Have Questions?
Catalog Number
28005
Unit Size
12 tests
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
2800512 tests
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Pierce Biotin Quantitation Kit includes aliquots of HABA reagent and biotinylated protein standard for convenient colorimetric determination of biotinylation levels in labeled antibodies and other proteins.

Features of the Biotin Quantitation Kit:

Colorimetric – requires a plate reader or spectrophotometer set to measure at 500nm
Reliable – proven, well-characterized method for determination of biotinylation levels
Convenient – kit provides the essential reagents and standards to perform the assay with ease
Flexible – adaptable to spectrophotometer cuvettes or standard plate readers with 96-well microplates
Options – calculate results directly from absorbance values based on extinction coefficients using the procedure outlined in the instructions
Robust – supplied HABA-avidin complex can be used over a wide range of pH and salt concentrations

HABA dye (4'-hydroxyazobenzene-2-carboxylic acid) is a reagent that enables quick estimation of the mole-to-mole ratio of biotin to protein in a solution. The Pierce Biotin Quantitation Kit contains a premix of HABA and avidin and a biotinylated horseradish peroxidase (HRP) positive control. The HABA-Avidin Premix is supplied in convenient Thermo Scientific No-Weigh Microtube packaging, which eliminates the difficulties associated with weighing small quantities of reagent.

To quantitate biotinylation, a solution containing the biotinylated protein is added to a mixture of HABA and avidin. Because of its higher affinity for avidin, biotin displaces the HABA and the absorbance at 500nm decreases proportionately. By this method, an unknown amount of biotin present in a solution can be quantitated in a single cuvette by measuring the absorbance of the HABA-avidin solution before and after addition of the biotin-containing sample. The change in absorbance relates to the amount of biotin in the sample by the extinction coefficient of the HABA-avidin complex. View online HABA Calculator.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
Biotin Quantitation Kit
Quantity
12 Tests
Product Line
Pierce™
Label or Dye
Biotin
Labeling Target
Antibodies, Proteins
Unit Size
12 tests

Contents & Storage

Sufficient For: Testing biotinylation of 12 protein samples
• HABA-Avidin Premix, 24 tubes
• Biotinylated Horseradish Peroxidase, 5 mg

Store at 4°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text