Thermo Scientific™
HABA (4'-hydroxyazobenzene-2-carboxylic acid)
Catalog number: 28010
Have Questions?
Thermo Scientific™
HABA (4'-hydroxyazobenzene-2-carboxylic acid)
Catalog number: 28010
Have Questions?
Catalog Number
28010
Unit Size
10 g
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
2801010 g
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Pierce HABA is 4'-hydroxyazobenzene-2-carboxylic acid, a simple reagent that enables spectrophotometric (colorimetric) estimation of biotinylation levels of labeled proteins and other molecules.
Features of HABA:
• HABA-avidin complex can be used over a wide range of pH and salt concentrations
• Amount of Avidin can be calculated directly from the increased absorbance at 500nm complexing with the HABA Dye
• Calculate results directly from absorbance values based on extinction coefficients using the procedure outlined in the instructions
• Complete kits also available! See Pierce Biotin Quantitation Kit (Part No. 28005) and Fluorescence Biotin Quantitation Kit (Part No. 46610)
Determine the molar ratio of biotin incorporated into a protein using the HABA-Avidin method. HABA dye (4'-hydroxyazobenzene-2-carboxylic acid ) binds to avidin to produce a yellow-orange colored complex which absorbs at 500nm. Free biotin will displace the HABA dye and cause the absorbance to decrease. A standard curve can be established using the free biotin to estimate the number of moles of biotin incorporated after biotinylating a protein. View online HABA Calculator.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product Type
HABA
Quantity
10 g
Product Line
Pierce™
Label or Dye
Biotin
Labeling Target
Antibodies, Proteins
Contents & Storage
Upon receipt store product at room temperature. Product shipped at ambient temperature.