Accessories for Luminoskan™ Ascent Microplate Luminometer
Accessories for Luminoskan™ Ascent Microplate Luminometer
Actual product may vary
Accessories for Luminoskan™ Ascent Microplate Luminometer
Accessories for Luminoskan™ Ascent Microplate Luminometer
Accessories for Luminoskan™ Ascent Microplate Luminometer
Run a variety of luminometric research applications with this compact, robust instrument certified as DLR-Ready.
Have Questions?
Catalog NumberProduct TypeNo. of Dispensers
2805621Dispenser Kit1
Catalog number 2805621
Price (USD)
20,400.00
Each
In stock
Add to cart
Product Type:
Dispenser Kit
No. of Dispensers:
1
Price (USD)
20,400.00
Each
Add to cart
Ask our AI about this Product
Run a variety of luminometric research applications with the Thermo Scientific™ Luminoskan™ Ascent Microplate Luminometer, a compact and robust instrument offering versatile plate formats, fast reading speeds and up to three dispensers. This instrument has passed the DLReady™ validation at Promega Corporation™ and is certified as DLReady. The Luminoskan Ascent Microplate Luminometer is validated for Thermo Scientific™ Pierce™ Luciferase Assays.
Fast reading speed:
  • Reads a 96-well plate in just 15 seconds
  • Essential for kinetic applications, such as enzyme kinetics and phagocytosis assays
Onboard dispensers for exact follow-up of kinetic reactions:
  • Equipped with up to three reagent dispensers for fast kinetic assays, such as ATP assays
  • Supports simultaneous dispensing and reading, enabling monitoring of fast kinetic measurements from the very start of the reaction
  • Offers very low dead volume and backflush capability for conservation of expensive reagents
High sensitivity:
  • DLReady certified for excellent sensitivity
  • Fiberless direct illumination optics ensures maximal light collection
  • High sensitivity, a wide dynamic range and low crosstalk for accurate and precise results in luminometric assays
Specially designed for automation:
  • Reads up to 384-well plates for increased assay throughput
  • Easily integrates with automated systems
Includes:
PC Software
Recommended for:
  • Reporter gene assays
  • Immunoassays with luminescent substrates
  • Cell proliferation and cytotoxicity assays
  • Intracellular Ca2+ assays
  • ATP assays
  • Phagocytosis assays
  • Reactive oxygen assays
  • Microbiological assays
  • Enzyme assays
  • BRET assays
  • ADMEtox
Specifications
CompatibilityRobot
No. of Dispensers1
Product TypeDispenser Kit
Plate Type384-well Plate
Product LineLuminoskan Ascent
Software TypePC Software
Unit SizeEach
Our new Varioskan ALF Multimode Microplate Reader for absorbance, fluorescence and luminescence measurements is fast, flexible & easy-to-use. We offer intuitive SkanIt software editions for Research or 21 CFR Part 11 compliance.
Learn more now
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Share catalog number, name or link
1x1 image pixel for data collection