1x1 image pixel for data collection
Triton™ X-100 Surfact-Amps™ Detergent Solution
Triton™ X-100 Surfact-Amps™ Detergent Solution
Thermo Scientific™

Triton™ X-100 Surfact-Amps™ Detergent Solution

Thermo Scientific Triton X-100 Surfact-Amps Detergent Solution is a highly-purified Triton™ X-100 detergent stabilized as a 10% solution (w/v) inRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
283146 x 10 mL
8511150 mL
85112250 mL
283131 L
Catalog number 28314
Price (USD)
156.65
Online Exclusive
181.00
Save 24.35 (13%)
Each
Add to cart
Quantity:
6 x 10 mL
Request bulk or custom format
Price (USD)
156.65
Online Exclusive
181.00
Save 24.35 (13%)
Each
Add to cart
Thermo Scientific Triton X-100 Surfact-Amps Detergent Solution is a highly-purified Triton™ X-100 detergent stabilized as a 10% solution (w/v) in variable sizes packaged under nitrogen in 10 mL glass ampules or three sizes of non-leaching HDPE bottles, ensuring their stability and eliminating the accumulation of peroxides and degradation products.

Features of Triton X-100 Surfact-Amps Detergent Solution:

Triton X-100—a nonionic detergent for use in various protein methods
Accurate—precise 10% detergent solution in ultrapure water
Easy-to-use—solution is simple to dispense and dilute for use
Exceptionally pure—less than 1.0μeq/mL peroxides and carbonyls
Stable—packaged under inert nitrogen gas in glass ampules or HDPE bottles

Specifications for Triton X-100 Surfact-Amps Detergent Solution:
• Visual: Clear to slightly hazy, colorless solution, free of particulate matter.
• Concentration: 10.0±1.0%
• Oxidants: ≤1.0μeq/mL
• Carbonyls: ≤1.0μeq/mL
• Suspended Solids: Residue present must not exceed Residue Reference.

Properties of Triton X-100:
• Molecular Weight: 647g
• Detergent Class: Nonionic polyoxyethylene surfactant
• Aggregation Number: 140
• Micelle Molecular Weight: 90,000g
• Critical Micelle Concentration (CMC): 0.24 mM (0.0155% w/v)
• Cloud Point: 64°C
• Dialyzable: No

Triton X-100 is most frequently used as a component of cell lysis buffers or other solutions intended to extract and solubilize proteins.

Related Products
Surfact-Amps™ Detergent Sampler
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity6 x 10 mL
Reagent TypeDetergent Solution
FormLiquid
Product LineSurfact-Amps™, Triton™
Product TypeDetergent
Unit SizeEach
Contents & Storage
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20°C to 25°C.
Learn about immunopeptidomics basics
Whether you're new to or experienced with proteomics, we're here to help.


Learn about immunopeptidomics basics ›
Follow 5-step sample prep workflow ›

Frequently asked questions (FAQs)

I need to do immunolabeling of a thick cryosection, around 60 µm thick. What should I do to ensure good labeling with my antibody?

One thing you should do is to permeabilize well, by including 0.1% to 0.2% Triton X-100 in all washes and incubations. Another recommendation is to extend the labeling times to overnight at 4oC for both primary antibody and secondary antibody. Finally, label the section free-floating, rather than adhered to a slide, to gain penetration for the antibody from both sides.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What are the storage conditions for Triton X-100 Surfact-Amps Detergent Solution (Cat. No. 85111)?

After opening, we recommend storing the unused solution sealed in an air-tight vial containing minimal air space or from which the air has been purged with nitrogen gas. Frequent opening should be avoided because Triton X-100 is susceptible to autoxidation upon exposure to air.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Can you provide the shelf life for Triton X-100 Surfact-Amps Detergent Solution (Cat. No. 85111)?

Triton X-100 Surfact-Amps Detergent Solution is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended (20-25 degrees C). Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection