NP-40 Surfact-Amps™ Detergent Solution
NP-40 Surfact-Amps™ Detergent Solution
Thermo Scientific™

NP-40 Surfact-Amps™ Detergent Solution

Thermo Scientific Surfact-Amps NP-40 Detergent Solution is highly-purified NP-40 detergent stabilized as a 10% solution in your choice of eitherRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
283246 x 10 mL
8512450 mL
85125500 mL
Catalog number 28324
Price (USD)
151.65
Online Exclusive
181.00
Save 29.35 (16%)
Each
In stock
Add to cart
Quantity:
6 x 10 mL
Request bulk or custom format
Price (USD)
151.65
Online Exclusive
181.00
Save 29.35 (16%)
Each
Add to cart
Thermo Scientific Surfact-Amps NP-40 Detergent Solution is highly-purified NP-40 detergent stabilized as a 10% solution in your choice of either 10 mL glass ampules or two sizes of HDPE-plastic bottles.

Features of Surfact-Amps NP-40 Detergent Solution:

NP-40—a nonionic detergent for use in various protein methods
Accurate—precise 10% detergent solutions in ultrapure water
Easy-to-use—solutions are simple to dispense and dilute for use
Exceptionally pure—less than 1.0μeq/mL peroxides and carbonyls
Stable—packaged under inert nitrogen gas in HDPE bottles

This is an easy-to-use 10% (w/v) solution of purified NP-40 detergent for use in routine and high-demand protein research methods and molecular biology techniques. NP-40 is a nonionic polyoxyethylene surfactant that is most frequently used as a component of cell lysis buffers or other solutions intended to extract and solubilize proteins. Surfact-Amps Detergent Solutions (10% w/v) provide unsurpassed purity, quality and stability. Unlike neat detergents, which are extremely viscous, Surfact-Amps 10% Solutions are easy to pipette and accurately dispense. The surfactant solutions are carefully prepared and packaged under nitrogen in glass ampules or non-leaching HDPE bottles, ensuring their stability and eliminating the accumulation of peroxides and degradation products.

Properties of NP-40 Detergent:
• Molecular Weight: 617g
• Detergent Class: Nonionic
• Aggregation Number: 149
• Micelle Molecular Weight: 90,000g
• Critical Micelle Concentration (CMC): 0.29 mM (0.0179%, w/v)
• Cloud Point: 80°C
• Dialyzable: No

Specifications for NP-40 Surfact-Amps Detergent Solution:
• Visual: Clear, colorless solution, free of particulate matter.
• Concentration: 10.0±1.0%
• Oxidants: ≤1.0μeq/mL
• Carbonyls: ≤1.0μeq/mL
• Suspended Solids: Residue present must not exceed Residue Reference.

Related Products
Surfact-Amps™ Detergent Sampler
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormatGlass Ampule
Quantity6 x 10 mL
Reagent TypeDetergent Solution
FormLiquid
Product LineSurfact-Amps™
Product TypeDetergent
Unit SizeEach
Contents & Storage
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20°C to 25°C.
Learn about immunopeptidomics basics
Whether you're new to or experienced with proteomics, we're here to help.


Learn about immunopeptidomics basics ›
Follow 5-step sample prep workflow ›

Frequently asked questions (FAQs)

Will NP-40 affect the migration of the samples in the SDS-PAGE gel?

Yes. All detergents and even phospholipids in cell extracts will form mixed micelles with SDS and migrate down into the gel.

They can also interfere with the SDS:protein binding equilibrium. Most of the nonionic detergents significantly interfere with SDS-PAGE.

We recommend that you keep the ratio of SDS to lipid or other detergent at 10:1 (or greater) to minimize these effects.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection