Thermo Scientific™

Brij™-58 Surfact-Amps™ Detergent Solution

Catalog number: 28336
Have Questions?
Thermo Scientific™

Brij™-58 Surfact-Amps™ Detergent Solution

Catalog number: 28336
Have Questions?
Catalog Number
28336
Unit Size
6 x 10 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
283366 x 10 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific™ Surfact-Amps™ 58 Detergent Solution is highly-purified Brij™ 58 detergent stabilized and supplied as a 10% solution (w/v) in 10 mL glass ampules.

Properties of Brij 58 Detergent:

• Molecular Weight: 1120g
• Detergent Class: Nonionic
• Aggregation Number: 70
• Micelle Molecular Weight: 82,000g
• Critical Micelle Concentration (CMC): 0.077mM (0.0086%, w/v)
• Cloud Point: >100°C

Features of Surfact-Amps 58 Detergent Solution:

Brij 58—a nonionic detergent for use in various protein methods
Accurate—10% detergent solutions in ultrapure water
Easy-to-use—solutions are simple to dispense and dilute for use

Specifications for Surfact-Amps 58 Detergent Solution:

• Visual: Clear to slightly hazy, colorless solution, free of particulates.
• Concentration: 10.0 ±1.0%
• Oxidants: ≤1.0µeq/mL
• Carbonyls: ≤1.0µeq/mL
• Suspended Solids: Residue present must not exceed Residue Reference.

For added safety when opening glass ampules, consider using our Ampule Breakers, which are disposable safety devices that protect the fingers when breaking open a glass ampule.

Related Products
Brij™-35 Surfact-Amps™ (10% w/v) Solution

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
Detergent
Form
Liquid
Product Line
Brij™, Surfact-Amps™
Reagent Type
Detergent Solution
Quantity
6 x 10 mL

Contents & Storage

Store at Room Temperature

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text