Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Concentration | Dry Blend |
Product Line | BupH™ |
Product Type | DPBS |
Quantity | 40 Packs |
pH | 7.4 |
Form | Powder |
Unit Size | 40 packs |
Concentration | Dry Blend |
Product Line | BupH™ |
Product Type | Phosphate Buffered Saline (PBS) |
Quantity | 40 Packs |
pH | 7.2 |
Form | Powder |
Unit Size | 40 packs |
Concentration | Dry Blend |
Product Line | BupH™ |
Product Type | MES Buffered Saline |
Quantity | 10 Packs |
For Use With (Application) | Carboxyl-to-Amine Crosslinking |
pH | 4.7 |
Reagent Type | MES Buffer |
Form | Powder |
Unit Size | 10 packs |
Product Line | BupH™ |
Product Type | Carbonate-Bicarbonate Buffer Packs |
Quantity | 40 Packs |
pH | 9.4 |
Form | Powder |
Unit Size | 40 packs |
Concentration | Dry Blend |
Product Line | BupH™ |
Product Type | Tris Buffered Saline Packs |
Quantity | 40 Packs |
For Use With (Application) | ELISA, Western Blotting |
pH | 7.2 |
Reagent Type | Tris Buffer |
Form | Powder |
Unit Size | 40 packs |
Concentration | Dry Blend |
Product Line | BupH™ |
Product Type | Borate Buffer |
Quantity | 40 Packs |
pH | 8.5 |
Form | Powder |
Unit Size | 40 packs |
Compatibility | SDS-PAGE Gels |
Concentration | Dry Blend |
Product Line | BupH™ |
Product Type | Tris-Glycine Buffer |
Quantity | 40 Packs |
For Use With (Application) | Wet Transfer |
Reagent Type | Transfer Buffer |
Form | Powder |
Unit Size | 40 packs |
Concentration | Dry Blend |
Product Line | BupH™ |
Product Type | Tris Buffered Saline Packs |
Quantity | 10 Packs |
For Use With (Application) | ELISA, Western Blotting |
pH | 7.2 |
Reagent Type | Tris Buffer |
Form | Powder |
Unit Size | 10 packs |
Catalog Number | Specifications | Unit Size | Product Type | Quantity | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
28374 | 40 packs | DPBS | 40 Packs | 198.65 Online price 213.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
28372 | 40 packs | Phosphate Buffered Saline (PBS) | 40 Packs | 195.65 Online price 212.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
28390 | 10 packs | MES Buffered Saline | 10 Packs | 163.65 Online price 177.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
28382 | 40 packs | Carbonate-Bicarbonate Buffer Packs | 40 Packs | 197.65 Online price 212.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
28376 | 40 packs | Tris Buffered Saline Packs | 40 Packs | 197.65 Online price 211.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
28384 | 40 packs | Borate Buffer | 40 Packs | 194.65 Online price 211.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
28380 | 40 packs | Tris-Glycine Buffer | 40 Packs | 161.65 Online price 175.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
28379 | 10 packs | Tris Buffered Saline Packs | 10 Packs | 93.65 Online price 102.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|