Invitrogen™

Dynabeads™ CaptureSelect™ AAVX Magnetic Beads

Catalog number: 2853522001
Have Questions?
Invitrogen™

Dynabeads™ CaptureSelect™ AAVX Magnetic Beads

Catalog number: 2853522001
Have Questions?
Catalog Number
2853522001
Unit Size
1 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
5 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
50 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
28535220011 mL
-
28535220055 mL
-
285352205050 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
DynaBeads CaptureSelect AAVX Magnetic Beads are high-capacity, high-throughput magnetic affinity particles for the purification of a broad range of naturally occurring and synthetic adeno-associated virus (AAV) serotypes used for gene therapy applications. The anti-AAVX affinity ligand demonstrates good binding reactivity towards a large variety of AAV serotypes, including AAV1 to AAV8, AAVrh10, and synthetic serotypes. The magnetic beads enable purification using manual and robotic magnetic separators of AAV serotypes directly from crude feedstock in a single step with high purity and yield.

Features of Dynabeads CaptureSelect AAVX Magnetic Beads include:
• One-magnetic-step AAV purification from crude material with high purity and yield
• Compatible with manual and automated workflows
• High specificity and capacity
• Easy upscaling to higher volumes
• Non-animal derived

Animal origin statement
CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a ∼12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics

Bead matrix: superparamagnetic
Bead size: 4.5 μm
Ligand: CaptureSelect AAVX affinity ligand
Binding capacity: ∼1 x1012 viral particles/mg beads
Elution conditions: 50 mM citric acid pH 2.5-3.0
Formulation buffer: tris-buffered saline, 0.05% Tween 20, 0.05 % sodium azide

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Binding Capacity
∼1 x 1012 viral particles/mg beads
Capacity (Metric)
1 mL
Concentration
30 mg beads/mL slurry, 1 mg contains approximately 1.3 x 107 beads
Formulation
Tris-buffered saline with 0.05% Tween-20 and 0.05% Sodium Azide
Matrix
Non-porous monodisperse superparamagnetic beads
Particle Size
4.5 μm
Form
Resin Slurry
Product Line
CaptureSelect™
For Use With (Application)
Magnetic isolation of AAV capsids
Target
Multiple AAV serotypes
Ligand Type
AAVX Affinity Ligand

Contents & Storage

Store at 2–8°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text