Thermo Scientific Chemicals

(2S,3R)-3-Phenylpyrrolidine-2-carboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: 290081000
100 mg, Glass bottle, Each
Thermo Scientific Chemicals

(2S,3R)-3-Phenylpyrrolidine-2-carboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: 290081000
100 mg, Glass bottle, Each
Quantity
Packaging
Catalog number: 290081000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
118758-48-8
IUPAC Name
(2S,3S)-3-phenylpyrrolidin-1-ium-2-carboxylate
Molecular Formula
C11H13NO2
InChI Key
VDEMEKSASUGYHM-UWVGGRQHSA-N
SMILES
[O-]C(=O)[C@H]1[NH2+]CC[C@H]1C1=CC=CC=C1
Appearance (Color)
White to off-white
Infrared spectrum
Conforms
Specific optical rotation
+82° to +86° (25°C, 589 nm) (c=0.75, 6 N HCl)
Appearance (Form)
Powder
HPLC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text