Thermo Scientific Chemicals

(2S,3S)-3-Methylpyrrolidine-2-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: 290090010
1 g, Glass Bottle, Each
Thermo Scientific Chemicals

(2S,3S)-3-Methylpyrrolidine-2-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: 290090010
1 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 290090010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10512-89-7
IUPAC Name
(2S,3S)-3-methylpyrrolidin-1-ium-2-carboxylate
Molecular Formula
C6H11NO2
InChI Key
CNPSFBUUYIVHAP-WHFBIAKZSA-N
SMILES
C[C@H]1CC[NH2+][C@@H]1C([O-])=O
Appearance (Form)
Crystalline powder
Appearance (Color)
White to beige
Infrared spectrum
Conforms
TLC
>=96.0 %
Specific optical rotation
-26° to -30° (25°C, 589 nm) (c=0.7, H2O)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text