1x1 image pixel for data collection
GC Liner Sealing Rings for Thermo Scientific™ and Agilent Instruments
GC Liner Sealing Rings for Thermo Scientific™ and Agilent Instruments
Thermo Scientific™
GC Liner Sealing Rings for Thermo Scientific™ and Agilent Instruments
Match the best sealing ring with the injection port liner used in your Thermo Scientific™ or Agilent GC system.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionFor Use With (Equipment)
29033406Liner Sealing RingTRACE and FOCUS GC SSL Injectors
290GA243Graphite Liner Sealing RingAgilent SSL Liners
290OA241Viton Liner Sealing RingTRACE 1300 Series GC and Agilent SSL Injectors
29001318TS
also known as 29001318
Liner Sealing RingTRACE 1300 Series GC PTV Injectors
29001320TSLiner Sealing RingTRACE 1300 Series GC SSL Injectors
Catalog number 29033406
Price (USD)
163.36
Pack of 10
Add to cart
Description:
Liner Sealing Ring
For Use With (Equipment):
TRACE and FOCUS GC SSL Injectors
Price (USD)
163.36
Pack of 10
Add to cart
Match the best sealing ring with the injection port liner used in your Thermo Scientific™ or Agilent GC system.
Specifications
DescriptionLiner Sealing Ring
For Use With (Equipment)TRACE and FOCUS GC SSL Injectors
Quantity10 Pack
Product TypeLiner Sealing Ring
Unit SizePack of 10
Share catalog number, name or link