1H-Benzimidazole-5-carboxylic acid, 98%, Thermo Scientific Chemicals
1H-Benzimidazole-5-carboxylic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1H-Benzimidazole-5-carboxylic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 290870250
Price (USD)/ Each
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
IUPAC Name1H-1,3-benzodiazole-6-carboxylic acid
Molecular FormulaC8H6N2O2
InChI KeyCOYPLDIXZODDDL-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C2N=CNC2=C1
Molecular Weight (g/mol)162.15
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with NaOH>=97.5 %
Appearance (Color)Grey to brown
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only