Triphenylphosphine dibromide, ca. 33% bromine, Thermo Scientific Chemicals
Triphenylphosphine dibromide, ca. 33% bromine, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Triphenylphosphine dibromide, ca. 33% bromine, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Catalog number 291070250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS1034-39-5
IUPAC Namedibromotriphenyl-λ⁵-phosphane
Molecular FormulaC18H15Br2P
InChI KeyOCXGTPDKNBIOTF-UHFFFAOYSA-N
SMILESBrP(Br)(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration Argentometricca. 33% Br2
Appearance (Color)Light yellow to light pink
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only