Thermo Scientific Chemicals

Triphenylphosphine dibromide, ca. 33% bromine, Thermo Scientific Chemicals

Catalog number: 291071000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Triphenylphosphine dibromide, ca. 33% bromine, Thermo Scientific Chemicals

Catalog number: 291071000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 291071000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1034-39-5
IUPAC Name
dibromotriphenyl-λ⁵-phosphane
Molecular Formula
C18H15Br2P
InChI Key
OCXGTPDKNBIOTF-UHFFFAOYSA-N
SMILES
BrP(Br)(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Form)
Crystalline powder
Titration Argentometric
ca. 33% Br2
Appearance (Color)
Light yellow to light pink
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text