Thermo Scientific Chemicals

Octyl 4-methoxycinnamate, 98%, stabilized, Thermo Scientific Chemicals

Catalog number: 291160250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Octyl 4-methoxycinnamate, 98%, stabilized, Thermo Scientific Chemicals

Catalog number: 291160250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 291160250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5466-77-3
IUPAC Name
2-ethylhexyl 3-(4-methoxyphenyl)prop-2-enoate
Molecular Formula
C18H26O3
InChI Key
YBGZDTIWKVFICR-UHFFFAOYNA-N
SMILES
CCCCC(CC)COC(=O)C=CC1=CC=C(OC)C=C1
Infrared spectrum
Conforms
Appearance (Form)
Liquid
Specific gravity
(25/25°C) 1.007 to 1.012
Stabilizer
0.05 to 0.1 % BHT
Appearance (Color)
Clear colorless to yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text