4,4'-Dichlorodiphenyl sulfone, 97%, Thermo Scientific Chemicals
4,4'-Dichlorodiphenyl sulfone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4,4'-Dichlorodiphenyl sulfone, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Packaging:
Plastic bottle
Catalog number 291180010
Price (USD)/ Each
130.00
-
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
130.00
Add to cart
4,4'-Dichlorodiphenyl sulfone, 97%, Thermo Scientific Chemicals
Catalog number291180010
Price (USD)/ Each
130.00
-
Add to cart
Chemical Identifiers
CAS80-07-9
IUPAC Name1-chloro-4-(4-chlorobenzenesulfonyl)benzene
Molecular FormulaC12H8Cl2O2S
InChI KeyGPAPPPVRLPGFEQ-UHFFFAOYSA-N
SMILESClC1=CC=C(C=C1)S(=O)(=O)C1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=96.0 %
Appearance (Form)Powder
Appearance (Color)White
Melting point146°C to 151°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only