Thermo Scientific Chemicals

4,4'-Dichlorodiphenyl sulfone, 97%, Thermo Scientific Chemicals

Catalog number: 291182500
250 g, Plastic bottle, Each
Thermo Scientific Chemicals

4,4'-Dichlorodiphenyl sulfone, 97%, Thermo Scientific Chemicals

Catalog number: 291182500
250 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 291182500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
80-07-9
IUPAC Name
1-chloro-4-(4-chlorobenzenesulfonyl)benzene
Molecular Formula
C12H8Cl2O2S
InChI Key
GPAPPPVRLPGFEQ-UHFFFAOYSA-N
SMILES
ClC1=CC=C(C=C1)S(=O)(=O)C1=CC=C(Cl)C=C1
Infrared spectrum
Conforms
GC
>=96.0 %
Appearance (Form)
Powder
Appearance (Color)
White
Melting point
146°C to 151°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text