Epithiochlorohydrine, 97%, Thermo Scientific Chemicals
Epithiochlorohydrine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Epithiochlorohydrine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 291390050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3221-15-6
IUPAC Name2-(chloromethyl)thiirane
Molecular FormulaC3H5ClS
InChI KeyXRWMHJJHPQTTLQ-UHFFFAOYNA-N
SMILESClCC1CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=96.0 %
Appearance (Form)Liquid
Refractive index1.5240 to 1.5280 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only