Thermo Scientific Chemicals

[1,3-Bis(diphenylphosphino)propane]nickel(II) chloride, 98+%, Thermo Scientific Chemicals

Catalog number: 291590010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

[1,3-Bis(diphenylphosphino)propane]nickel(II) chloride, 98+%, Thermo Scientific Chemicals

Catalog number: 291590010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 291590010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
15629-92-2
IUPAC Name
[3-(diphenylphosphanyl)propyl]diphenylphosphane; dichloronickelbis(ylium)
Molecular Formula
C27H26Cl2NiP2
InChI Key
REQVOKPBMSCTNI-UHFFFAOYSA-L
SMILES
Cl[Ni++]Cl.C(CP(C1=CC=CC=C1)C1=CC=CC=C1)CP(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
Red to red-brown
Appearance (Form)
Powder or crystals
Infrared spectrum
Conforms
Elemental Analysis
>=98.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text