Thermo Scientific Chemicals

[2,2]-Paracyclophane, 99%, Thermo Scientific Chemicals

Catalog number: 291600250
25 g, Each
Thermo Scientific Chemicals

[2,2]-Paracyclophane, 99%, Thermo Scientific Chemicals

Catalog number: 291600250
25 g, Each
Quantity
Catalog number: 291600250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1633-22-3
IUPAC Name
tricyclo[8.2.2.2⁴,⁷]hexadeca-1(12),4,6,10,13,15-hexaene
Molecular Formula
C16H16
InChI Key
OOLUVSIJOMLOCB-UHFFFAOYSA-N
SMILES
C1CC2=CC=C(CCC3=CC=C1C=C3)C=C2
HPLC
>=98.5 %
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder or crystals
Melting point
285°C to 288°C
Appearance (Color)
White to off-white

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text