Thermo Scientific Chemicals

(Aminomethyl)phosphonic acid, 99%, Thermo Scientific Chemicals

Catalog number: 291642500
250 mg, Glass bottle, Each
Thermo Scientific Chemicals

(Aminomethyl)phosphonic acid, 99%, Thermo Scientific Chemicals

Catalog number: 291642500
250 mg, Glass bottle, Each
Quantity
Packaging
Catalog number: 291642500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1066-51-9
IUPAC Name
(aminomethyl)phosphonic acid
Molecular Formula
CH6NO3P
InChI Key
MGRVRXRGTBOSHW-UHFFFAOYSA-N
SMILES
NCP(O)(O)=O
Titration with NaOH
>=98.5 %
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text