Thermo Scientific Chemicals

Phenylsilane, 97+%, Thermo Scientific Chemicals

Catalog number: 291690050
5 mL, Glass bottle, Each
Thermo Scientific Chemicals

Phenylsilane, 97+%, Thermo Scientific Chemicals

Catalog number: 291690050
5 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 291690050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
694-53-1
IUPAC Name
phenylsilyl
Molecular Formula
C6H7Si
InChI Key
JAZKZOWBOMBYIX-UHFFFAOYSA-N
SMILES
[SiH2]C1=CC=CC=C1
Refractive index
1.5085 to 1.5135 (20°C, 589 nm)
GC
>=97.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text