Thermo Scientific Chemicals

Potassium sorbate, 99%, Thermo Scientific Chemicals

Catalog number: 291741000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Potassium sorbate, 99%, Thermo Scientific Chemicals

Catalog number: 291741000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 291741000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
24634-61-5
IUPAC Name
potassium (2E,4E)-hexa-2,4-dienoate
Molecular Formula
C6H7KO2
InChI Key
CHHHXKFHOYLYRE-STWYSWDKSA-M
SMILES
[K+].C\C=C\C=C\C([O-])=O
Appearance (Color)
White to cream
Appearance (Form)
Powder or granules
Infrared spectrum
Conforms
Titration with HClO4
98.5 to 101.5 %
Loss on drying
=<1 % (105°C, 3 h)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text