1-Cyclohexene-1-carboxylic acid, 96%, Thermo Scientific Chemicals
1-Cyclohexene-1-carboxylic acid, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Cyclohexene-1-carboxylic acid, 96%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
2.5 g
Packaging:
Glass bottle
Catalog number 291750025
Price (USD)/ Each
-
Quantity:
2.5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namecyclohex-1-ene-1-carboxylic acid
Molecular FormulaC7H10O2
InChI KeyNMEZJSDUZQOPFE-UHFFFAOYSA-N
SMILESOC(=O)C1=CCCCC1
Molecular Weight (g/mol)126.16
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline mass or liquid
HPLC>=95 %
Appearance (Color)White to light yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only