Thermo Scientific Chemicals

Sodium triacetoxyborohydride, 97%, Thermo Scientific Chemicals

Catalog number: 291825000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Sodium triacetoxyborohydride, 97%, Thermo Scientific Chemicals

Catalog number: 291825000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 291825000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
56553-60-7
IUPAC Name
sodium bis(acetyloxy)boranuidyl acetate
Molecular Formula
C6H10BNaO6
InChI Key
HHYFEYBWNZJVFQ-UHFFFAOYSA-N
SMILES
[Na+].CC(=O)O[BH-](OC(C)=O)OC(C)=O
Appearance (Form)
Powder
Infrared spectrum
Conforms
Appearance (Color)
White
Titration with HClO4
96.0 to 104.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text