Thermo Scientific Chemicals

Dicobalt octacarbonyl, 95%, stabilized, Thermo Scientific Chemicals

Catalog number: 291840050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Dicobalt octacarbonyl, 95%, stabilized, Thermo Scientific Chemicals

Catalog number: 291840050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 291840050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10210-68-1
Molecular Formula
C8Co2O8
InChI Key
ZUDHYLQHRXCZGU-UHFFFAOYSA-N
SMILES
[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].O=[C-]1[Co][C-](=O)[Co++]1
Molecular Weight (g/mol)
341.95
Assay
>=94 %
Appearance (Form)
Crystals or flakes
Stabilizer
=<5 % hexane
Appearance (Color)
Orange-red to dark red
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text