Dicobalt octacarbonyl, 95%, stabilized, Thermo Scientific Chemicals
Dicobalt octacarbonyl, 95%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dicobalt octacarbonyl, 95%, stabilized, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
View all
Glass bottle
Catalog number 291840250
Price (USD)/ Each
233.00
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
233.00
Add to cart
Dicobalt octacarbonyl, 95%, stabilized, Thermo Scientific Chemicals
Catalog number291840250
Price (USD)/ Each
233.00
-
Add to cart
Chemical Identifiers
CAS10210-68-1
Molecular FormulaC8Co2O8
InChI KeyZUDHYLQHRXCZGU-UHFFFAOYSA-N
SMILES[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].O=[C-]1[Co][C-](=O)[Co++]1
Molecular Weight (g/mol)341.95
SpecificationsSpecification SheetSpecification Sheet
Assay>=94 %
Appearance (Form)Crystals or flakes
Stabilizer=<5 % hexane
Appearance (Color)Orange-red to dark red
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only