Chemical Identifiers
CAS32005-36-0
IUPAC Namebis((1E,4E)-1,5-diphenylpenta-1,4-dien-3-one) palladium
Molecular FormulaC34H28O2Pd
InChI KeyUKSZBOKPHAQOMP-SVLSSHOZSA-N
SMILES[Pd].O=C(\C=C\C1=CC=CC=C1)/C=C/C1=CC=CC=C1.O=C(\C=C\C1=CC=CC=C1)/C=C/C1=CC=CC=C1
View more