Pierce™ NeutrAvidin™ Agarose
Pierce™ NeutrAvidin™ Agarose
Thermo Scientific™

Pierce™ NeutrAvidin™ Agarose

Thermo Scientific NeutrAvidin Agarose is a standard-capacity beaded agarose resin of immobilized NeutrAvidin Protein, a modified form of avidin withRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
2920110 mL
292005 mL
Catalog number 29201
Price (USD)/ 10 mL
557.65
Online exclusive
586.00 
Save 28.35 (5%)
-
Add to cart
Quantity:
10 mL
Request bulk or custom format
Price (USD)/ 10 mL
557.65
Online exclusive
586.00 
Save 28.35 (5%)
Add to cart
Pierce™ NeutrAvidin™ Agarose
Catalog number29201
Price (USD)/ 10 mL
557.65
Online exclusive
586.00 
Save 28.35 (5%)
-
Add to cart
Thermo Scientific NeutrAvidin Agarose is a standard-capacity beaded agarose resin of immobilized NeutrAvidin Protein, a modified form of avidin with exceptional biotin-binding characteristics for affinity purification methods.

Features of NeutrAvidin Agarose:

NeutrAvidin protein—purified, deglycosylated avidin protein (60kDa, pI 6.3); tetrameric with four biotin-binding site per molecule
Agarose resin—support is crosslinked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods
Inert and stable—NeutrAvidin protein is immobilized by charge-free, leach-resistant covalent bonds, resulting in low nonspecific binding and enabling multiple uses without decline in yield
Standard capacity—this variety of beads has a normal load of immobilized NeutrAvidin protein, providing a binding capacity of 1 to 2 mg of biotinylated BSA per mL of resin

NeutrAvidin Agarose consists of a specially purified and deglycosylated form of avidin that has been covalently immobilized onto high-quality crosslinked 6% beaded agarose. Compared to native avidin, NeutrAvidin protein is more neutral and exhibits much less nonspecific binding properties in biotin-binding applications. The agarose beads have physical and chemical properties that enable them to be used in a variety of batch- or column-type affinity procedures. Generally, NeutrAvidin products function as well or better than their streptavidin counterparts and are less expensive.

NeutrAvidin protein is deglycosylated native avidin from egg whites. Removal of the excess carbohydrate by an exclusive process yields a protein with a more neutral isoelectric point and less nonspecific binding properties. NeutrAvidin resins are prepared by covalently coupling the protein using efficient and stable chemistries, resulting in supports that are resistant to leaching and stable at pH 2-11. The products are excellent choices for a variety of small- or large-scale affinity purification applications involving biotinylated macromolecules, including separation of biotinylated molecules from samples and immunoprecipitation of antigens using biotin-labeled antibodies.

Properties of crosslinked 6% beaded agarose (CL-6B):
• Support pH Stability: 2 to 14 (short term); 3 to 13 (long term)
• Average Particle Size: 45 to 165 microns
• Exclusion Limit: 10,000 to 4,000,000 daltons
• Maximum Volumetric Flow Rate: approx. 1 mL/minute (for 1 cm diameter column)
• Maximum Linear Velocity: 30 cm per hour
• Maximum Pressure: less than 25psi (1.5 bar)

Related Products
Pierce™ High Capacity NeutrAvidin™ Agarose
Pierce™ NeutrAvidin™ UltraLink™ Resin
Pierce™ NeutrAvidin™ Plus UltraLink™ Resin
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Particle Size45 to 165 μm
Product LineNeutrAvidin™, Pierce™
Quantity10 mL
TargetBiotin-Tagged Molecule
TypeAgarose
Stationary PhaseNeutrAvidin Protein
FormatBottle
Column TypeAffinity, Agarose Resin
Unit Size10 mL
Contents & Storage
Upon receipt store at 4°C. Do not freeze resin. Product is shipped at ambient temperature.