Thermo Scientific™

Pierce™ High Capacity NeutrAvidin™ Agarose

Catalog number: 29204
Have Questions?
Thermo Scientific™

Pierce™ High Capacity NeutrAvidin™ Agarose

Catalog number: 29204
Have Questions?
Catalog Number
29204
Unit Size
10 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
5 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
2920410 mL
-
292025 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific High Capacity NeutrAvidin Agarose is a beaded agarose resin of immobilized NeutrAvidin Protein, a modified form of avidin with exceptional biotin-binding characteristics for affinity purification methods.

Features of High Capacity NeutrAvidin Agarose:

NeutrAvidin protein—purified, deglycosylated avidin protein (60kDa, pI 6.3); tetrameric with four biotin-binding site per molecule
Agarose resin—support is crosslinked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods
Inert and stable—NeutrAvidin protein is immobilized by charge-free, leach-resistant covalent bonds, resulting in low nonspecific binding and enabling multiple uses without decline in yield
High capacity—this variety of beads has a dense load of immobilized NeutrAvidin protein, providing a binding capacity greater than 8 mg of biotinylated BSA per mL of resin

High Capacity NeutrAvidin Agarose consists of a specially purified and deglycosylated form of avidin that has been covalently immobilized at a high density onto high-quality crosslinked 6% beaded agarose. Compared to native avidin, NeutrAvidin protein is more neutral and exhibits much less nonspecific binding properties in biotin-binding applications. The agarose beads have physical and chemical properties that enable them to be used in a variety of batch- or column-type affinity procedures. Generally, NeutrAvidin products function as well or better than their streptavidin counterparts and are less expensive.

NeutrAvidin protein is deglycosylated native avidin from egg whites. Removal of the excess carbohydrate by an exclusive process yields a protein with a more neutral isoelectric point and less nonspecific binding properties. NeutrAvidin resins are prepared by covalently coupling the protein using efficient and stable chemistries, resulting in supports that are resistant to leaching and stable at pH 2-11. The products are excellent choices for a variety of small- or large-scale affinity purification applications involving biotinylated macromolecules, including separation of biotinylated molecules from samples and immunoprecipitation of antigens using biotin-labeled antibodies.

Properties of crosslinked 6% beaded agarose (CL-6B):
• Support pH Stability: 2 to 14 (short term); 3 to 13 (long term)
• Average Particle Size: 45 to 165 microns
• Exclusion Limit: 10,000 to 4,000,000 daltons
• Maximum Volumetric Flow Rate: approx. 1 mL/minute (for 1 cm diameter column)
• Maximum Linear Velocity: 30 cm per hour
• Maximum Pressure: less than 25psi (1.5 bar)

Related Products
Pierce™ NeutrAvidin™ Agarose
Pierce™ NeutrAvidin™ UltraLink™ Resin
Pierce™ NeutrAvidin™ Plus UltraLink™ Resin
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Capacity (Metric)
8 mg Biotinylated BSA Binding per mL Resin
Description
Pierce High Capacity NeutrAvidin Agarose
Particle Size
45 to 165 μm
Product Line
NeutrAvidin™, Pierce™
Quantity
10 mL
Target
Biotin-Tagged Molecule
Type
Agarose
Stationary Phase
NeutrAvidin Protein
Format
Bottle
Column Type
Affinity, Agarose Resin

Contents & Storage

Upon receipt store at 4°C. Do not freeze resin. Product is shipped at ambient temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text