Thermo Scientific Chemicals

Isobornyl methacrylate, 85-90%, stabilized, Thermo Scientific Chemicals

Catalog number: 292092500
250 g, Glass bottle, Each
Thermo Scientific Chemicals

Isobornyl methacrylate, 85-90%, stabilized, Thermo Scientific Chemicals

Catalog number: 292092500
250 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 292092500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7534-94-3
IUPAC Name
1,7,7-trimethylbicyclo[2.2.1]heptan-2-yl 2-methylprop-2-enoate
Molecular Formula
C14H22O2
InChI Key
IAXXETNIOYFMLW-UHFFFAOYNA-N
SMILES
CC(=C)C(=O)OC1CC2CCC1(C)C2(C)C
Appearance (Color)
Clear colorless to yellow
Refractive index
1.4750 to 1.4790 (20°C, 589 nm)
Appearance (Form)
Liquid
Stabilizer
100 to 200 ppm MEHQ
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text