o-Tolyl isocyanate, 99+%, Thermo Scientific Chemicals
o-Tolyl isocyanate, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

o-Tolyl isocyanate, 99+%, Thermo Scientific Chemicals

Have Questions?
Quantity:
10 g
Packaging:
Glass bottle
Catalog number 292220100
Price (USD)/ Each
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS614-68-6
IUPAC Name1-isocyanato-2-methylbenzene
Molecular FormulaC8H7NO
InChI KeyVAYMIYBJLRRIFR-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1N=C=O
View more
SpecificationsSpecification SheetSpecification Sheet
AppearanceMay develop some turbidity or precipitate
GC>=99.0 %
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only