Thermo Scientific Chemicals

2-Chlorophenyl isothiocyanate, 99%, Thermo Scientific Chemicals

Catalog number: 292240050
5 g, Each
Thermo Scientific Chemicals

2-Chlorophenyl isothiocyanate, 99%, Thermo Scientific Chemicals

Catalog number: 292240050
5 g, Each
Quantity
Catalog number: 292240050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2740-81-0
IUPAC Name
1-chloro-2-isothiocyanatobenzene
Molecular Formula
C7H4ClNS
InChI Key
DASSPOJBUMBXLU-UHFFFAOYSA-N
SMILES
ClC1=CC=CC=C1N=C=S
Appearance (Form)
Liquid
GC
>=98.5 %
Appearance (Color)
Clear yellow
Infrared spectrum
Conforms
Refractive index
1.6598 to 1.6630 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text