Gibco™

Phytone™ Peptone

Catalog number: 292450
Have Questions?
Gibco™

Phytone™ Peptone

Catalog number: 292450
Have Questions?
Catalog Number
292450
Unit Size
10 kg
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
454 g
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
29245010 kg
-
211906454 g
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Phytone Peptone is an animal origin-free (AOF) enzymatic digest of soy. Due to the naturally high concentrations of vitamins and carbohydrates in soy, Phytone Peptone is a nutritious and excellent carbohydrate source. Customers have successfully used this supplement in CHO-based production of monoclonal antibodies for oncological and rheumatological therapeutics, as well as with many different microbial cell types to produce animal health vaccines for swine, such as Clostridial vaccines.

Features:
• Animal origin-free (AOF) supplement
• Enzymatic digests of soybean meal/flour
• Retains the high vitamin and high carbohydrate content of the soy plant tissue

Applications of Phytone Peptone include:
• It is an excellent plant peptone for the cultivation of fungi and fastidious types of bacteria, such as members of the Clostridium and Neisseria genera
• It has been used in mammalian cell culture applications due to its high carbohydrate content

Specifications

Form
Powder
Product Type
Peptone
For Use With (Application)
Production of monoclonal antibodies for oncological and rheumatological therapeutics, microbial cell types to produce animal health vaccines for swine, such as Clostridial vaccines
Product Line
Phytone™
Quantity
10 kg
Source
Animal origin-free, soy
Unit Size
10 kg

Contents & Storage

Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text