Thermo Scientific Chemicals

Petroleum ether, extra pure, boiling range 60-95°C, Thermo Scientific Chemicals

Catalog number: 292510025
2.5 L, Glass Bottle, Each
Thermo Scientific Chemicals

Petroleum ether, extra pure, boiling range 60-95°C, Thermo Scientific Chemicals

Catalog number: 292510025
2.5 L, Glass Bottle, Each
Quantity
Packaging
Catalog number: 292510025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
64742-49-0
IUPAC Name
Petroleum ether
MDL Number
MFCD00081849
Refractive index
1.3880 to 1.3920 (20°C, 589 nm)
Color scale
=<10 APHA
Boiling point
60°C to 95°C
Aromatics
=<0.01 %
n-Hexane
=<2 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text