Thermo Scientific Chemicals

Methacrolein, 90%, stabilized, Thermo Scientific Chemicals

Catalog number: 292831000
100 mL, Glass bottle, Each
Thermo Scientific Chemicals

Methacrolein, 90%, stabilized, Thermo Scientific Chemicals

Catalog number: 292831000
100 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 292831000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
78-85-3
IUPAC Name
2-methylprop-2-enal
Molecular Formula
C4H6O
InChI Key
STNJBCKSHOAVAJ-UHFFFAOYSA-N
SMILES
CC(=C)C=O
GC
>=88 %
Appearance (Color)
Clear colorless to light yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Stabilizer
0.1 to 0.5 % Hydroquinone

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text