Methacrolein, 90%, stabilized, Thermo Scientific Chemicals
Methacrolein, 90%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methacrolein, 90%, stabilized, Thermo Scientific Chemicals

Methacrolein, 90%, C4H6O, CAS Number-78-85-3, 2-methyl-2-propenal, 2-methylpropenal, 2-methylacrolein, 2-methylenepropanal, 2-propenal, 2-methyl, isobutenal, methacrylaldehyde, methacrylic aldehyde, methylacrylaldehyde, methacrolein, 50mL, 88% min. (GC), 01, III, 2981, 69.0 deg.C
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
10 mL
50 mL
100 mL
Packaging:
Glass bottle
Catalog number 292835000
Price (USD)/ Each
-
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS78-85-3
IUPAC Name2-methylprop-2-enal
Molecular FormulaC4H6O
InChI KeySTNJBCKSHOAVAJ-UHFFFAOYSA-N
SMILESCC(=C)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=88 %
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Stabilizer0.1 to 0.5 % Hydroquinone
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only