Thermo Scientific Chemicals

6-Methylbenzoxazole, 99.5+%, Thermo Scientific Chemicals

Catalog number: 293110025
2.5 g, Glass bottle, Each
Thermo Scientific Chemicals

6-Methylbenzoxazole, 99.5+%, Thermo Scientific Chemicals

Catalog number: 293110025
2.5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 293110025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10531-80-3
IUPAC Name
6-methyl-1,3-benzoxazole
Molecular Formula
C8H7NO
InChI Key
SZWNDAUMBWLYOQ-UHFFFAOYSA-N
SMILES
CC1=CC=C2N=COC2=C1
Melting point
29°C to 31°C
Appearance (Color)
Amber
Infrared spectrum
Conforms
Appearance (Form)
Low melting solid
GC
>=99.50 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text