1,1,1,3,3,3-Hexafluoro-2-propanol, 99.9%, for spectroscopy, Thermo Scientific Chemicals
1,1,1,3,3,3-Hexafluoro-2-propanol, 99.9%, for spectroscopy, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,1,1,3,3,3-Hexafluoro-2-propanol, 99.9%, for spectroscopy, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 293410100
Price (USD)/ Each
118.65
Online exclusive
132.00 
Save 13.35 (10%)
-
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
118.65
Online exclusive
132.00 
Save 13.35 (10%)
Add to cart
1,1,1,3,3,3-Hexafluoro-2-propanol, 99.9%, for spectroscopy, Thermo Scientific Chemicals
Catalog number293410100
Price (USD)/ Each
118.65
Online exclusive
132.00 
Save 13.35 (10%)
-
Add to cart
Chemical Identifiers
CAS920-66-1
IUPAC Name1,1,1,3,3,3-hexafluoropropan-2-ol
Molecular FormulaC3H2F6O
InChI KeyBYEAHWXPCBROCE-UHFFFAOYSA-N
SMILESOC(C(F)(F)F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
UVat 195 nm A: =<0.097
Residue after evaporation=<0.001 %
Appearance (Form)Clear liquid
GC>=99.85 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopy
RUO – Research Use Only