Thermo Scientific Chemicals

1,1,1,3,3,3-Hexafluoro-2-propanol, 99.9%, for spectroscopy, Thermo Scientific Chemicals

Catalog number: 293410500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

1,1,1,3,3,3-Hexafluoro-2-propanol, 99.9%, for spectroscopy, Thermo Scientific Chemicals

Catalog number: 293410500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 293410500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
920-66-1
IUPAC Name
1,1,1,3,3,3-hexafluoropropan-2-ol
Molecular Formula
C3H2F6O
InChI Key
BYEAHWXPCBROCE-UHFFFAOYSA-N
SMILES
OC(C(F)(F)F)C(F)(F)F
Color scale
=<10 APHA
UV
at 195 nm A: =<0.097
Residue after evaporation
=<0.001 %
Appearance (Form)
Clear liquid
GC
>=99.85 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopy
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text